Learn More
Eppendorf™ Rotor T-60-11 with Lid
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
5810/5810R Centrifuge
Unit Size:
Each
Description
- Rotor T-60-11
- For Centrifuge 5810/5810 R
- Includes rotor lid
- For 1.5/2.0mL tubes
- Without adapter
Specifications
Specifications
| Type | Rotor T-60-11 with Lid |
| For Use With (Equipment) | 5810/5810R Centrifuge |
| Includes | Lid |