Learn More
BUCHI Lyovapor™ Drying Chamber
Product Code. 18804171 Shop All Buchi Products
Click to view available options
Type:
Benchtop Freeze Dryer
Heated Product Shelf Chamber
No. of Shelves:
0
4
6
Style:
12-position Manifold Top
24-position Manifold Top
Standard Top
Stoppering Vials Top
Includes:
4 Sample Temperature Sensors
6 Sample Temperature Sensors
Chamber Only
Drying Chamber Only
Pressure Difference Testing
Unit Size:
Each
Product Code. 18804171
Brand: BUCHI 11DCPH24101
Freeze drying flexibility for basic and advanced drying of vials, shelves, and flasks.
  • Manifolds: adapters for traditional freeze drying beakers or round bottom flasks
  • Shelves: for vials, plates, and irregular shaped items
  • Heated Shelves: advanced control of drying process and applications
  • Stoppering Chambers: seal vials without exposure to ambient moisture
Specifications
Type Heated Product Shelf Chamber
No. of Shelves 4
Temperature Range (Metric) Ambient to -105°C
Material Acrylic
No. of Ports 4
Style Standard Top
Quantity Heated Product Shelf Chamber
Includes Pressure Difference Testing
Plug Type North America
Voltage 220/240 V
Shipping Weight (English) 220/240 V