Learn More
IKA™ DT-20-M-Gamma Tube with Rotor Stator Element
Product Code. 12814831 Shop All IKA Products
Click to view available options
For Use With (Equipment):
DT-20-M
Unit Size:
Each
Product Code. 12814831
Brand: IKA™ 0003700600
Only null left
Specifications
Product Type DT-20-M Tube
Volume Range (Metric) 5 to 15 mL
For Use With (Application) Dispersion, homogenization, suspensions
For Use With (Equipment) DT-20-M
Packaging Quantity 20 Pk.