Learn More
Fisherbrand™ Anodized Dual Aluminum Blocks
Click to view available options
Product Type:
D1196, 96-Well
D12384, 384 WELL PCR PLATE
Unit Size:
Each
Description
- Excellent heat transfer
- Anodized to resist thermal shock
- For holding test tubes for digital heaters
- Available to fit a range of tube counts and sizes
Specifications
Specifications
| For Use With (Equipment) | PCR |
| Product Type | D12384, 384 WELL PCR PLATE |
| Quantity | 1 CT |
| Weight | 1 kg |