Learn More
Scientific Industries SI™ Dual Microplate Foam Insert
Click to view available options
No. of Positions:
2
4 (2 inserts)
Unit Size:
Each
Description
- Accommodates two standard footprint microplates.
- For use with Vortex-Genie™ MAX
Specifications
Specifications
| Material | Foam |
| For Use With (Equipment) | Vortex-Genie™ MAX |
| No. of Positions | 4 (2 inserts) |