Learn More
Thermo Scientific™ Dual QuantStudio System Integration Kit for the Orbitor™ RS2 Microplate Mover and QuantStudio™ Real-time PCR Systems
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Description
- This Dual QuantStudio System Integration Kit is designed for use with the Orbitor RS2 Microplate Mover and identical QuantStudio Systems, enabling real-time PCR in automated environments for increased sample throughput.
- This integration kit includes a location bracket, splitter, and Thermo Scientific Momentum Workflow Software for the second QuantStudio integration.
- Couple the Dual QuantStudio Integration Kit with the rbitor RS2 Microplate Mover for QuantStudio Systems and one of the two plate storage and handling options (Sequential Access or Random Access ) to complete your automation bundle.
Specifications
Specifications
| For Use With (Equipment) | QuantStudio 7 Pro, QuantStudio 7 Flex, or QuantStudio 12 K Flex Real-Time PCR Systems and The Orbitor RS2 Microplate Mover |
| Product Type | Integration Kit |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Room Temperature |
| Quantity | 1 kit |