missing translation for 'onlineSavingsMsg'
Learn More

Dual Standard Solution for YSI2300 Glucose/Lactate Analyzer, Reagecon

Product Code. 11563202 Shop All Reagecon Products
Click to view available options
Quantity:
250 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11563202

Brand: Reagecon Y2747

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dual Standard Solution for YSI2300 Glucose/Lactate Analyzer

  • Dual Standard Solution for YSI2300 Glucose/Lactate Analyzer.

Specifications

Packaging HDPE Bottle
Physical Form Liquid
For Use With (Equipment) YSI2300 Glucose/Lactate Analyzer
Quantity 250 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.