Learn More
Thermo Scientific Chemicals Duloxetine hydrochloride
Product Code. p-7214443 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16671822
Brand: Thermo Scientific Chemicals 463140010
Request Bulk or Custom Format
333.87 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Molecular Weight (g/mol) 333.87
Product Type Duloxetine hydrochloride
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol
compliance-icons
Product Identifier
  • Duloxetine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Duloxetine hydrochloride