missing translation for 'onlineSavingsMsg'
Learn More

Duloxetine Phenyl Carbamate (>85%), TRC

Product Code. 30136333
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136333 100 mg 100mg
17964895 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136333 Supplier Toronto Research Chemicals Supplier No. TRCD721030100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947686-09-1
Formula Weight 417.1399
InChI Formula InChI=1S/C25H23NO3S/c1-26(25(27)28-20-11-3-2-4-12-20)17-16-23(24-15-8-18-30-24)29-22-14-7-10-19-9-5-6-13-21(19)22/h2-15,18,23H,16-17H2,1H3/t23-/m0/s1
IUPAC Name phenyl N-methyl-N-[(3S)-3-naphthalen-1-yloxy-3-thiophen-2-ylpropyl]carbamate
Molecular Formula C25H23NO3S
Molecular Weight (g/mol) 417.52
SMILES Oc1ccc(cc1)/C=C/C(=O)Nc2c(cccc2)CCC3N(C)CCCC3
Synonym N-Methyl-N-[(3S)-3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]carbamic Acid Phenyl Ester, (S)-Duloxetine Impurity A;
Chemical Name or Material Duloxetine Phenyl Carbamate

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.