Learn More
Euro Protection™ Dunlop™ Devon S5 Safety Boots
Product Code. 12024465
Click to view available options
Size:
10
39
40
45
Unit Size:
Each
Product Code. 12024465
Brand: Euro Protection™ 9DESA45
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Certifications/Compliance CE Certified, EN ISO 20345:2011
Color Green, Black, White
Footbed Material Rubber-Nitrile
Heel Style Flat
Height (Metric) Heel 19.05 mm
Liner Material Polyester
Material Nitrile
Size 45
Sole Features Black color, anti-slip sole
Sole Material Rubber-Nitrile
Style Safety
Toe Material Steel
Resistant To Acids, various chemical products, toecap very resistant to shocks and squashing
Waterproof Yes
Product Type Safety Boot