Learn More
DWK Life Sciences DURAN™ Adapter, stopper type
Compact reduction from sleeve to core.
Brand: DWK Life Sciences 286576503
Additional Details : Weight : 0.02000kg
Description
DURAN™ adapters are used to connect two components with different NS sizes. For the reduction from a larger core to a smaller core with low space requirements.
- Compact reduction from sleeve to core.
- Due to its compact design, this transition piece takes up very little space.
- Stopper shape
Specifications
10/Pk. | |
Connection of two components with different standard ground joint sizes | |
Solid | |
Socket NS 14/23, Cone NS 29/32 |