Learn More
DWK Life Sciences DURAN™ Blank for separating and dropping funnel, cylindrical with stopper bed for ISO 4800
Raw body for further processing as dropping funnel.
Brand: DWK Life Sciences 210144405
249.85 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 2.30000kg
Description
DURAN™ dropping funnels are used, for example, for the metered supply of liquid to a reaction mixture.
- Cylindrical with stopper bed for ISO 4800
- Glass articles for further processing
Specifications
65 mm | |
Boro 3.3 | |
Blank for further processing as dropping funnel; Phase separation |
500 mL | |
Yes | |
10/Pk. |