Learn More
Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Angled
Product Code. 10375211
Click to view available options
Capacity (Metric):
25 mL
100 mL
250 mL
500 mL
1000 mL
5 L
Socket Joint:
14/23 NS Side Neck, 14/23 NS Center Neck
14/23 NS Side Neck, 24/29 NS Center Neck
14/23 NS Side Neck, 29/32 NS Center Neck
19/26 NS Side Neck, 24/29 NS Center Neck
19/26 NS Side Neck, 29/32 NS Center Neck
29/32 NS Side Neck, 29/32 NS Center Neck
Unit Size:
Each
Product Code. 10375211
Brand: Lenz Laborglasinstrumente™ 03314237
Only null left
Add to basket
Add to basket
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Angled are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN 12394
  • Two side necks at 20° angles
  • With conical ground joints, featuring high taper accuracy and excellent tightness
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass, Duran Glass
Socket Joint 19/26 NS Side Neck, 29/32 NS Center Neck
Bottom Shape Round