missing translation for 'onlineSavingsMsg'
Learn More

SCAT DURAN™ Bottles, Round

Laboratory bottles made of high quality and shatterproof DURAN glass.

Brand:  SCAT 501121

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 14707260

  • 76.90€ / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

  • Screw caps with GL45 or GLS80 threads
  • Made of clear or amber glass
  • Round bottles
  • All common sizes from 100 ml to 10000 ml volume
  • Optionally with Protect coating or wide neck opening
  • DURAN™ Glass from the German manufacturer Schott for the highest demands in the laboratory

    Specifications

    Specifications

    Safety Waste Caps
    Brown
    HPLC Systems
    GL 45
    DURAN Glass
    Not Autoclavable
    Round
    250 mL
    Laboratory bottle DURAN, GL45, 250 mL, brown glass & round, with cap and pouring ring
    Cap and pouring ring
    Blue
    Yes
    Reusable
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Promotions

    Promotions