missing translation for 'onlineSavingsMsg'
Learn More

SCAT DURAN™ Bottles, Square

Product Code. 14737250 Shop All SCAT Europe Products
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 14737250

Brand: SCAT 501112

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Laboratory bottles made of high quality and shatterproof DURAN glass.

  • Screw caps with GL45 threads
  • Made of clear or amber glass
  • Available as round or square bottles
  • All common sizes from 250 mL to 1000 mL volume
  • Optionally with Protect coating or wide neck opening
  • DURAN™ Glass from the German manufacturer Schott for the highest demands in the laboratory

    Specifications

    Capacity (Metric) 250 mL
    Capacity (English) 0.054 gal.
    Color Clear
    Description Laboratory bottle DURAN, GL45, 250 mL, clear glass & square , with cap and pouring ring
    For Use With (Application) HPLC Systems
    Material Borosilicate Glass
    Sterility Non-sterile
    Thread Size (Metric) GL 45
    Closure Color Blue
    Closure Included Yes
    Disposable Reusable
    Graduated Yes
    Mouth Narrow
    Shape Square
    Product Type Square Glass Bottle
    For Use With (Equipment) Safety Waste Caps
    Autoclavable Not Autoclavable
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.