Learn More
DWK Life Sciences DURAN™ Connection Piece, with standard ground cone, 90° angle
The connector allows the connection of equipment via a hose.
Brand:  DWK Life Sciences 243000801
18.00 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.01800kg
 View more versions of this product
Product Code. 10203701
  • 18.95€ / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
Connectors are transition pieces with olive and have an olive instead of the second standard ground joint. Tubes can be connected via this olive, which allow e.g. the connection of a vacuum pump.
  • Tight closure due to standard ground joint
  • 90° angle
  • With standard ground cone
  • Easy and tight connection of hoses
  • Fired-on, highly durable white print
Specifications
Specifications
1 Ea.
Connection of hoses
NS 24/29