Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Adapter
Duran™ Glass Adapter made of DURAN™ tubing.
44.73€ - 54.75€
Specifications
Cone Size | NS 29/32 |
---|---|
Material | Duran Glass |
Product Type | Adapter |
Quantity | 1 Ea. |
Product Code | Brand | Cone Size | Standard Taper Joint No. | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Standard Taper Joint No. | Price | Quantity & Availability | ||||
10101761
|
Lenz Laborglasinstrumente™
01523529 |
NS 29/32 | NS 35 |
54.75€
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
10141711
|
Lenz Laborglasinstrumente™
01521929 |
NS 29/32 | NS 19 |
44.73€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10294381
|
Lenz Laborglasinstrumente™
01532935 |
NS 29/32 | NS 29 |
53.21€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10335831
|
Lenz Laborglasinstrumente™
01532929 |
NS 29/32 | NS 29 |
46.27€
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
Description
- Duran™ Glass
Specifications
NS 29/32 | |
Duran Glass | |
Adapter | |
1 Ea. |