Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Screw Thread Tube
Duran™ Glass Screw Thread Tube with NS cone, cap, and sealing.
23.80€ - 40.23€
Specifications
Includes | Screw-cap aperture, Silicone sealing ring, PTFE washers |
---|---|
Material | Duran Glass |
Product Type | Screw thread Tube |
Quantity | 1 Ea. |
Product Code | Brand | Cone Size | Thread Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Thread Size | Price | Quantity & Availability | ||||
11913341
|
Lenz Laborglasinstrumente™
01301814 |
NS 14/23 | GL 18 |
23.80€
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
11923341
|
Lenz Laborglasinstrumente™
01301819 |
NS 19/26 | GL 18 |
28.20€
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||||
11903341
|
Lenz Laborglasinstrumente™
01301429 |
NS 29/32 | GL 14 |
33.69€
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
11953341
|
Lenz Laborglasinstrumente™
01302529 |
NS 29/32 | GL 25 |
40.23€
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
Description
- Duran™ Glass
Specifications
Screw-cap aperture, Silicone sealing ring, PTFE washers | |
Duran Glass | |
Screw thread Tube | |
1 Ea. |