Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Socket Adapter
Brand:  Lenz Laborglasinstrumente™ 01100129
Additional Details : Weight : 0.05600kg
 View more versions of this product
Product Code. 10412931
  • 38.27€ / Pack of 10
Estimated Shipment: 15-07-2024
to see stock.
Specifications
DIN 12249 form E and in addition to DIN standard
130mm
Socket Adapter
Socket Size/Cone Size: NS 29/32
Duran Glass
10Pack