missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Glass Stopper, Clear, Semi-hollow, Standard Taper Ground Joint, Octagonal flat head

Clear DURAN™ glass ground stopper.

Brand:  DWK Life Sciences 216241209

17.58 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.



Additional Details : Weight : 0.14800kg

 View more versions of this product

Product Code. 10315351

  • 18.50€ / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

Semi-hollow or solid DURAN™ glass stoppers can be used to close glassware items such as flasks, separatory funnels or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!

  • All DURAN™ glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to Standard Taper ground joint
  • Octagonal shaped head
Specifications

Specifications

Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders
Boro 3.3
No
1 Ea.
ST 45/40
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions