Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, flat, diamond-ground
Hollow glass stopper for sealing.
Brand:  DWK Life Sciences 286121808
Additional Details : Weight : 12.03200kg
 View more versions of this product
Product Code. 15931521
  • 268.00€ / Pack of 100
Estimated Shipment: 16-08-2023
to see stock.
Add to basket
Description
Description
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
  • Flat
  • Diamond-ground
  • hollow glass stopper
Specifications
Specifications
Closure of vessels with standard ground joints
100/Pk.
Yes
NS 7/16
Boro 3.3