Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground
Hollow glass stopper for sealing with draining tip.
Brand: DWK Life Sciences 286118103
Additional Details : Weight : 0.38000kg
Description
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
- Pointed
- Diamond-ground
- With a dripping syringe, for example, a condensate can be collected and the reaction medium recirculated.
- hollow glass stopper
Specifications
Closure of vessels with standard ground joints | |
10/Pk. | |
Yes |
NS 45/40 | |
Boro 3.3 |