Learn More
DWK Life Sciences DURAN™ Micro Filter Candle, without tube
The micro filter candle is suitable for the extraction of supernatant solutions but also for the separation of solids from a carrier gas.
396.00€ - 447.00€
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Clarification filtration; Extraction of clear filtrate (not the filtrand); Separation of solids from carrier gas; Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering |
No. per Pack | 10/Pk. |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15282059
|
DWK Life Sciences
258572107 |
Porosity 1 |
396.00€
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
15292059
|
DWK Life Sciences
258572201 |
Porosity 2 |
396.00€
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
15212069
|
DWK Life Sciences
258572407 |
Porosity 4 |
447.00€
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11650029
|
DWK Life Sciences
258572004 |
Porosity 0 |
396.00€
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
15202069
|
DWK Life Sciences
258572304 |
Porosity 3 |
396.00€
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
Description
One of the most important separation methods in the laboratory environment is filtration, which is suitable for the separation or purification of different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Due to their size, microfilter candles are used for the filtration of very small quantities. This filtration is possible from a liquid or a carrier gas.
- Without tube
- Micro filter candle without tube can be used as a filter to separate the smallest amount of solids from the filter surface
Specifications
Borosilicate Glass (3.3) | |
Clarification filtration; Extraction of clear filtrate (not the filtrand); Separation of solids from carrier gas; Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering | |
10/Pk. |