Learn More
3M™ Dustmaster™ Particulate Filters
Particulate filter for use with the Dustmaster Powered Air Respirator helps protects against dust, mist and fume.
Brand:  3M™ 7000059865
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11751363
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Combines with the 6000 Series Gas Vapor cartridges to provide particulate and gas/vapor protection
  • Note: 3M 501 filter retainers needed to combine the 5000 and 6000 series
Specifications
Specifications
P2
6000 and 7000 series facemasks
5/Pk.
Dust Filter
Filter