missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ DVB/Amidine Latex Beads, 4% w/v, 0.5μm

Product Code. 11580306
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Product Code. Quantity unitSize
11580306 15 mL 15mL
1 options
This item is not returnable. View return policy

Product Code. 11580306

Brand: Molecular Probes™ D37472

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

DVB/Amidine Latex Beads, 4% w/v, 0.5 μm

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.

The CML/sulfonate microspheres are negatively charged polystyrene microspheres with sulfonate and high density of carboxyl functional groups on the surface. The surface charge is pH dependent and hydrophilic in nature.

Microspheres Are Available in Many Sizes and Surface Functionalities
  • 25 sizes to choose from—0.02 microns to 16 microns
  • 20 different surface—for ready conjugation to virtually any material
  • Surfactant free—ultraclean
  • Various quantities—from milliliter to 500-liter amounts

    Key Applications of Microspheres
  • Instrument calibration (flow cytometry, microscopy, HTS, HCS)
  • Flow testing (microfluidics, blood flow, water flow, and air flow)
  • Cell biology tracers (cell differentiation and cell tracing)
  • Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

    Get More Information
    Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.

    We’ll Make a Custom Microsphere Product for You
    If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 9001:2000 certified.

    For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 0.04w/v
    Content And Storage 2°C to 8°C
    Description DVB/Amidine Latex Beads, 15mL, 4% w/v Concentration, 0.5μm Particle Size, White, Suspension Form, De-ionized Water Medium, Specialty Latex Beads, Room Temperature, 2 to 8°C Storage Temperature, Ideal Materials for the Adsorption of Materials Such a
    Color White
    For Use With (Application) Instrument Calibration (Flow Cytometry, Microscopy, HTS, HCS), Flow Testing (Microfluidics, Blood Flow, Water Flow, and Air Flow), Cell Biology Tracers (Cell Differentiation and Cell Tracing), Immunoassays (Agglutination Tests, ELISA, Particle Capture, and Contrast Reagents)
    Material Polystyrene
    Format Suspension
    Packaging 1 Each
    Parking Area 85 A2/SO4 (Amidine Group)
    Particle Density 1.055g/cm3 at 20°C
    Quantity 15 mL
    Refractive Index 1.591 at 20°C
    Surface Modification DVB-Amidine
    Volume (Metric) 15 mL
    Diameter (Metric) Mean 0.5 μm
    Diameter (Metric) 0.5 μm
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.