Learn More
Cole-Parmer™ Dye Concentrate
Product Code. 11777947 Shop All Cole-Parmer Products
Change view
Click to view available options
Color:
Blue
Dark Red
Yellow/Green
pH Range:
5.1 to 5.3
8
8.6 to 8.8
Unit Size:
Each
Product Code. Color pH Range unitSize
11777947 Dark Red 8.6 to 8.8 Each
11797947 Blue 5.1 to 5.3 Each
11787947 Yellow/Green 8 Each
3 options
Product Code. 11777947
Brand: Cole-Parmer™ 0029816
Determine water flow in closed circuits or open channels with these dyes. Dye Concentrate diffuses rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, flow rates, discharge locations, seepage areas, sea marking and more.
  • Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters.
  • Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult
  • Won’t harm the environment—nontoxic and biodegradable
  • Easy-to-use color in rapidly diffusing dye identifies dispersion patterns, flow rates, seepage and more Noncarcinogenic fluorescent red dye is ideal for yellowed or heavily algaed water
  • Brighten murky ponds with blue dye
  • Minimum detectable concentration is 1 ppm
Specifications
Color Dark Red
pH Range 8.6 to 8.8
For Use With (Application) Squirt-top dispensing
Quantity 1/Pk.
Type Dye Tracer