Description
Cole-Parmer Dye Tablets, Yellow/Green, Dye Tracer, For Use With: Plumbing, storm and sanitary drain analysis, Quantity: 200/Pk.
Specifications
Specifications
| Color | Yellow/Green |
| For Use With (Application) | Plumbing, storm and sanitary drain analysis |
| Quantity | 200/Pk. |
| Type | Dye Tracer |