missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ CD8

Product Code. 30275206
Click to view available options
Quantity:
2 mL
5 mL
Unit Size:
2mL
Each
This item is not returnable. View return policy

Product Code. 30275206

Brand: Invitrogen™ 11148D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dynabeads CD8 are superparamagnetic beads coupled with an anti-human CD8 antibody that enable easy isolation or depletion of human CD8+ T cells directly from any sample, including whole blood, bone marrow, buffy coat, PBMC and tissue digests.

Dynabeads CD8 are superparamagnetic beads coupled with an anti-human CD8 antibody that enable easy isolation or depletion of human CD8+ T cells directly from any sample, including whole blood, bone marrow, buffy coat, peripheral blood mononuclear cells (PBMCs), and tissue digests. Advantages of Dynabeads CD8:

  • Fast and efficient isolation of CD8+ T cells from any sample—no columns required
  • High purity, yield, and viability of positively isolated CD8+ T cells for molecular downstream assays
  • Fast and efficient depletion of CD8+ T cells from any sample

About Dynabeads CD8

Dynabeads CD8 are uniform, superparamagnetic beads (4.5 μm diameter) coated with a primary monoclonal antibody specific for the CD8 membrane antigen mainly expressed on human cytotoxic T cells. Because of the bead size, Dynabeads CD8 can easily and efficiently isolate or deplete cells from viscous samples such as whole blood and bone marrow in about 30 minutes. Dynabeads CD8 also afford excellent recovery of high-purity, viable cells for use in downstream molecular studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, so if you want to isolate CD8+ cells for cell-based applications, or you need to check your samples with flow cytometry, check out our total human CD8 product offerings.

Magnetic bead to based separation offers easy handling

Dynabeads CD8 is added to the sample under continuous mixing to optimize the binding of the Dynabeads to the target cells. By placing the sample on a magnet, the bead-bound target cells are separated from the rest of the sample in just 1 to 2 minutes. For depletion, remove the supernatant to a new tube for further studies and discard the bead-bound cells. For positive isolation for molecular studies, remove the supernatant and wash the bead-bound cells 2 to 3 times in buffer to get optimal purity. The cells can be lysed while still attached to the beads and the supernatant transferred to a new tube for downstream molecular analysis. Starting samples can be whole blood, buffy coat, bone marrow, PBMC, or tissue digests.

Automation-ready for use with KingFisher purification systems

The product is ideal for high-throughput workflows using KingFisher systems. A user guide and scripts for KingFisher Flex and Apex workflows are available for download on this page.

Learn more about Dynabeads products

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells
Content And Storage This Product Contains: 2 mL Dynabeads™ CD8 coated with anti-CD8 monoclonal antibody.
Storage Conditions: 2°C to 8°C. Do not freeze.
Color Brown
For Use With (Application) Cell Depletion, Positive Isolation for Molecular Applications
Material Magnetic Polystyrene
High-throughput Compatibility Yes
No. of Cells Processes ∼8x 10^8 cells total
Output Viability >95%
Product Line Dynabeads
Purity or Quality Grade Research Grade
Quantity 2 mL
Sample Type PBMC, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 10^7 PBMCs per isolation
Target Species Human
Diameter (Metric) 4.5 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.