Learn More
Abeba™ Dynamic 37315 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
66.43€
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | Black |
Material | Leather |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15640878
|
Abeba™
37315/35 |
35 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15650878
|
Abeba™
37315/36 |
36 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15660878
|
Abeba™
37315/37 |
37 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15670878
|
Abeba™
37315/38 |
38 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15680878
|
Abeba™
37315/39 |
39 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15690878
|
Abeba™
37315/40 |
40 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15600888
|
Abeba™
37315/41 |
41 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15610888
|
Abeba™
37315/42 |
42 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15620888
|
Abeba™
37315/43 |
43 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15630888
|
Abeba™
37315/44 |
44 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15640888
|
Abeba™
37315/45 |
45 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15650888
|
Abeba™
37315/46 |
46 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15660888
|
Abeba™
37315/47 |
47 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15670888
|
Abeba™
37315/48 |
48 |
66.43€
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather
- Black/brown
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Polyurethane |
Black | |
Slip-resistant | |
37315 |