Learn More
Abeba™ Dynamic 37331 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
69.04€
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | Black |
Material | Microfiber |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15680888
|
Abeba™
37331/35 |
35 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15690888
|
Abeba™
37331/36 |
36 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15600898
|
Abeba™
37331/37 |
37 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15610898
|
Abeba™
37331/38 |
38 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15620898
|
Abeba™
37331/39 |
39 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15630898
|
Abeba™
37331/40 |
40 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15640898
|
Abeba™
37331/41 |
41 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15650898
|
Abeba™
37331/42 |
42 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15660898
|
Abeba™
37331/43 |
43 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15670898
|
Abeba™
37331/44 |
44 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15680898
|
Abeba™
37331/45 |
45 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15690898
|
Abeba™
37331/46 |
46 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15600908
|
Abeba™
37331/47 |
47 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15610908
|
Abeba™
37331/48 |
48 |
69.04€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Microfiber black/blue
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Microfiber | |
Polyurethane |
Black | |
Slip-resistant | |
37331 |