Learn More
Abeba™ Dynamic 37333 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand: Abeba™ 37333/41
Additional Details : Weight : 0.98000kg
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Microfiber black/yellow
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Seamles upper
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Microfiber | |
Slip-resistant | |
37333 |
Black | |
41 | |
Polyurethane |