Learn More
Abeba™ Dynamic 37345 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
72.95€
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | White, Black |
Material | Microfiber |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15660988
|
Abeba™
37345/35 |
35 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15670988
|
Abeba™
37345/36 |
36 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15680988
|
Abeba™
37345/37 |
37 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15690988
|
Abeba™
37345/38 |
38 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15600998
|
Abeba™
37345/39 |
39 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15610998
|
Abeba™
37345/40 |
40 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15620998
|
Abeba™
37345/41 |
41 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15630998
|
Abeba™
37345/42 |
42 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15640998
|
Abeba™
37345/43 |
43 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15650998
|
Abeba™
37345/44 |
44 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15660998
|
Abeba™
37345/45 |
45 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15670998
|
Abeba™
37345/46 |
46 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15680998
|
Abeba™
37345/47 |
47 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15690998
|
Abeba™
37345/48 |
48 |
72.95€
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Microfiber with breathable textile inlays
- White/Black
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Microfiber | |
Polyurethane |
White, Black | |
Slip-resistant | |
37345 |