Learn More
Abeba™ Dynamic 37360 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
71.53€
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | White |
Material | Microfiber |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15601008
|
Abeba™
37360/35 |
35 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15611008
|
Abeba™
37360/36 |
36 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15621008
|
Abeba™
37360/37 |
37 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15631008
|
Abeba™
37360/38 |
38 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15641008
|
Abeba™
37360/39 |
39 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15651008
|
Abeba™
37360/40 |
40 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15661008
|
Abeba™
37360/41 |
41 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15671008
|
Abeba™
37360/42 |
42 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15681008
|
Abeba™
37360/43 |
43 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15691008
|
Abeba™
37360/44 |
44 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15601018
|
Abeba™
37360/45 |
45 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15611018
|
Abeba™
37360/46 |
46 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15621018
|
Abeba™
37360/47 |
47 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15631018
|
Abeba™
37360/48 |
48 |
71.53€
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Microfiber
- White
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Microfiber | |
Polyurethane |
White | |
Slip-resistant | |
37360 |