Learn More
Abeba™ Dynamic 37363 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 37363/39
Additional Details : Poids : 0.98000kg
 View more versions of this product
Product Code. 15661048
  • 48.40€ / 1 pair
Expédition estimée
29-09-2020
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber
  • Gray/Red
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
37363
Gray, Red
39
Polyurethane