Learn More
Abeba™ Dynamic 7331 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Brand:  Abeba™ 7331/41
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 15632698
  • 66.43€ / 1 pair
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber black/blue
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Seamless upper
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
7331
Black
41
Polyurethane