Learn More
Abeba™ Dynamic 7345 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Brand:  Abeba™ 7345/43
Additional Details : Poids : 0.99000kg
 View more versions of this product
Product Code. 15652768
  • 59.07€ / 1 pair
Expédition estimée
12-02-2021
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber with breathable textile inlays
  • White/Black
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
7345
White, Black
43
Polyurethane