missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Neuritin Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
Neuritin protein is fully biologically active when compared to standard. The ED50 as determined by by a cell proliferation assay using rat C6 cells is less than 25 ng/mL, corresponding to a specific activity of >4.0 x 10^4 IU/mg..
Specifications
Specifications
| Accession Number | Q9NPD7, Q9NPD7, Q9NPD7 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 51299 |
| Molecular Weight (g/mol) | 19.4 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of Neuritin as determined by LAL method. |
| Gene Alias | dJ380B8.2, neuritin, neuritin 1, NRNMGC44811 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction