Learn More
Invitrogen™ E-Gel™ Agarose Gels Starter Pak, 0.8% (EU only)
Click to view available options
Quantity:
Each
Unit Size:
Each
Description
E-Gel™ Single Comb Gels are supplied in a 12-well mini-gel format, with a choice of either SYBR Safe™, or ethidium bromide DNA stain. Each gel contains a single row of 12 sample wells. E-Gels are available in two pack sizes-a Starter Pak, and 18-gel multi-paks.
Specifications
Specifications
| Description | E-Gel™ Agarose Gels Starter Pak, 0.8%, E-Gel™ Gel Type, Ethidium Bromide In-Gel Dye, 12 Wells, Single Comb Well Format, Store at Room Temperature, For Nucleic Acid Gel Electrophoresis and Blotting |
| Concentration | 0.8% |
| Product Type | Agarose Gels Starter Kit |
| For Use With (Application) | DNA and RNA purification analysis, nucleic acid gel electrophoresis and blotting, agarose gel electrophoresis |
| Kit Contents | 6 E-Gels, the E-Gel™ PowerBase™ v.4, and a manual, 18-packs contain 18 gels and a manual |
| Quantity | Each |
| Storage Requirements | Room temperature |