missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ EX Agarose Gels Basic Starter Kit, 2%, EU Version

Product Code. 15491134
Change view
Click to view available options
Quantity:
Each
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491134 Each 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15491134 Supplier Invitrogen™ Supplier No. G6412STEU

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

E-Gel EX Agarose Gels Basic Starter Kit, 2%, EU Version

The E-Gel™ EX 2% Agarose Basic Starter Kit is an economical way to get started using E-Gel™ EX agarose gels. It provides:

  • One box of 10 E-Gel™ EX 2% Agarose Gels
  • E-Gel™ iBase™ Power System
  • E-Gel™ 1 Kb Plus DNA Ladder
  • E-Gel™ Sample Loading Buffer
  • One gel knife

    E-Gel™ EX 1% Agarose Gels are self-contained, pre-cast agarose gels designed to provide fast, convenient, and easy electrophoresis. Each E-Gel™ agarose gel is ready for use with agarose, electrodes, and SYBR™ Gold DNA gel stain packaged inside an 11-well, easy-to-open, UV- and blue-light transparent cassette. SYBR™ Gold stain can be viewed with blue light, which minimizes DNA damage and allows real-time monitoring of DNA migration with the E-Gel™ Safe Imager Real-Time Transilluminator.

    Ultra-sensitive detection of DNA
    E-Gel™ EX gels were developed for ultimate sensitivity and demonstrate over 5-fold greater sensitivity than comparable gels containing ethidium bromide. This is due to the use of the ultra-sensitive SYBR™ Gold nucleic acid gel stain. The superior sensitivity allows you to use lower amounts of sample, saving time and money.

    Quick check of RNA samples
    E-Gel™ EX gels can also be used for fast analysis of RNA samples to check on their integrity before proceeding with downstream applications.

    Easy access to the gel
    The gel cassette is designed to open with a gel knife (included with this kit), so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis.

    Running and viewing apparatus
    E-Gel™ EX gels require the E-Gel™ iBase™ Power System (included) for running and the E-Gel™ Safe Imager™ Real-Time Transilluminator (not included) for real-time viewing of sample migration.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Description E-Gel™ EX Agarose Gels Basic Starter Kit, 2%, E-Gel™ Gel Type, EU Version, For Electrophoresis, For Use With Agarose, Electrodes, and SYBR™ Gold DNA Gel Stain Packaged Inside an 11-Well, UV- and Blue-Light Transparent Cassette
    Concentration 2%
    For Use With (Application) DNA and RNA purification analysis, nucleic acid gel electrophoresis and blotting, agarose gel electrophoresis
    Kit Contents One box of 10 E-Gel™ EX 2% agarose gels, E-Gel™ iBase™ power system, E-Gel™ 1 Kb plus DNA ladder, E-Gel™ sample loading buffer, one gel knife
    Quantity Each
    Product Type Agarose Gels Starter Kit

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.