Learn More
Abeba™ Easy 37622 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca: Abeba™ 37622/36
Dettagli aggiuntivi : Weight : 0.97000kg
Descrizione
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather white/blue
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3573)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
Specifica
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Slip-resistant | |
37622 |
White | |
36 | |
Polyurethane |