Learn More
Abeba™ Easy 37622 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 37622/41
Additional Details : Weight : 0.87300kg
 View more versions of this product
Product Code. 15691098
  • 67.85€ / 1 pair
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white/blue
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
37622
White
41
Polyurethane