Learn More
Abeba™ Easy 37630 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
63.95€
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRC
Color White
Material Leather
Sole Features Slip-resistant
Sole Material Polyurethane
View More Specs
Products 14
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15661118
View Documents Promotion Details
Abeba™
37630/35
35
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15671118
View Documents Promotion Details
Abeba™
37630/36
36
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15681118
View Documents Promotion Details
Abeba™
37630/37
37
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15691118
View Documents Promotion Details
Abeba™
37630/38
38
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15601128
View Documents Promotion Details
Abeba™
37630/39
39
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15611128
View Documents Promotion Details
Abeba™
37630/40
40
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15621128
View Documents Promotion Details
Abeba™
37630/41
41
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15631128
View Documents Promotion Details
Abeba™
37630/42
42
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15641128
View Documents Promotion Details
Abeba™
37630/43
43
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15651128
View Documents Promotion Details
Abeba™
37630/44
44
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15661128
View Documents Promotion Details
Abeba™
37630/45
45
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15671128
View Documents Promotion Details
Abeba™
37630/46
46
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15681128
View Documents Promotion Details
Abeba™
37630/47
47
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15691128
View Documents Promotion Details
Abeba™
37630/48
48
63.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather, perforated
  • White
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Polyurethane
White
Slip-resistant
37630