Learn More
Abeba™ Easy 7621 Shoes
Includes TPU Sole with high slip resistance. Upper leather and microfiber construction.
Brand:  Abeba™ 7621/43
Additional Details : Poids : 0.97000kg
 View more versions of this product
Product Code. 15662878
  • 47.85€ / 1 pair
Expédition estimée
02-10-2020
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber
  • Patent Black
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
7621
Patent Black
43
Polyurethane