Learn More
Abeba™ Easy 7630 Shoes
Includes TPU Sole with high slip resistance. Upper leather and microfiber construction.
Brand:  Abeba™ 7630/39
Additional Details : Weight : 0.97000kg
 View more versions of this product
Product Code. 15642908
  • 61.34€ / 1 pair
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather, perforated
  • White
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
7630
White
39
Polyurethane