Learn More
Abeba™ Easy 7631 Shoes
Includes TPU Sole with high slip resistance. Upper leather and microfiber construction.
Brand: Abeba™ 7631/46
Additional Details : Poids : 0.97000kg
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather, perforated
- Black
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3573)
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Slip-resistant | |
7631 |
Black | |
46 | |
Polyurethane |