Learn More
Abeba™ Easy 7631 Shoes
Includes TPU Sole with high slip resistance. Upper leather and microfiber construction.
Brand:  Abeba™ 7631/36
Additional Details : Weight : 0.97000kg
 View more versions of this product
Product Code. 15652918
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather, perforated
  • Black
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
7631
Black
36
Polyurethane