Learn More
Greiner Bio-One Easyload Pipette Tip Refill System, Sterile
Product Code. 11709482
Click to view available options
Volume (Metric):
200 μL
1000 μL
Unit Size:
Pack of 2160
Pack of 5760
Product Code. 11709482
Brand: Greiner Bio-One 741061
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Stacked refill system with rack suitable for refilling pipette tip boxes.
  • Simple, time- and space-saving refill system for standard pipette tips.
  • Autoclavable racks.
  • Stackable racks.
  • Color-coded boxes.
  • Positioning guides make it easier to place refill units on the racks.
  • Design of rack interior increases stability during autoclaving and when pipette tips are bein used with multi-channel pipettes.
  • Graduated
  • Sterile
  • For single use only
  • Polypropylene construction
  • Free of heavy metal
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 μL
Sterility Sterile
Autoclavable Autoclavable
Sterilization Method Irradiation
Packaging 960/Pk
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Suitable for single-channel Brand, Eppendorf, Finnpipette, Gilson, Socorex pipettes, multi-channel Brand, Gilson, Eppendorf, Socorex pipettes
Graduated Yes
Product Line EasyLoad
Material Polypropylene