Learn More
Greiner Bio-One Easyload Pipette Tip Refill System, Sterile
Stacked refill system with rack suitable for refilling pipette tip boxes.
Brand: Greiner Bio-One 741061
Additional Details : Weight : 0.80000kg
Description
- Simple, time- and space-saving refill system for standard pipette tips.
- Autoclavable racks.
- Stackable racks.
- Color-coded boxes.
- Positioning guides make it easier to place refill units on the racks.
- Design of rack interior increases stability during autoclaving and when pipette tips are bein used with multi-channel pipettes.
- Graduated
- Sterile
- For single use only
- Polypropylene construction
- Free of heavy metal
Specifications
200 μL | |
Autoclavable | |
960/Pk | |
Single-use | |
Yes | |
Polypropylene |
Sterile | |
Irradiation | |
Yellow | |
Suitable for single-channel Brand, Eppendorf, Finnpipette, Gilson, Socorex pipettes, multi-channel Brand, Gilson, Eppendorf, Socorex pipettes | |
EasyLoad |