missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Easyload Pipette Tip Refill System, Sterile

Product Code. 11769472
Click to view available options
Volume (Metric):
200 μL
1000 μL
Unit Size:
Pack of 2160
Pack of 5760
This item is not returnable. View return policy

Product Code. 11769472

Brand: Greiner Bio-One 741040

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Stacked refill system with rack suitable for refilling pipette tip boxes.

  • Simple, time- and space-saving refill system for standard pipette tips.
  • Autoclavable racks.
  • Stackable racks.
  • Color-coded boxes.
  • Positioning guides make it easier to place refill units on the racks.
  • Design of rack interior increases stability during autoclaving and when pipette tips are bein used with multi-channel pipettes.
  • Graduated
  • Sterile
  • For single use only
  • Polypropylene construction
  • Free of heavy metal
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Sterility Sterile
Autoclavable Autoclavable
Sterilization Method Irradiation
Packaging 360/Pk
Color Code Blue
Disposable Single-use
For Use With (Equipment) Suitable for single-channel Brand Eppendorf Finnpipette, Gilson, Socorex pipettes
Graduated Yes
Product Line EasyLoad
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.