missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Direct HLA T Cell Isolation Kit
Click to view available options
Quantity:
100 mL whole blood
Unit Size:
1 set
Description
- The EasySep™ Direct HLA T Cell Isolation Kit can be used to isolate highly purified T cells directly from human whole blood, buffy coat, spleen, or lymph nodes by immunomagnetic negative selection.
- This kit targets non-T cells for removal with antibodies recognizing specific surface markers.
- Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet.
- Desired cells are simply collected into a new tube and can directly be used for downstream applications such as flow cytometry crossmatch.
Specifications
Specifications
| Cell Type | T Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 100 mL whole blood |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction